Learn More
Biomatik Corporation Dog High mobility group protein B1 (HMGB1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC32055
Encompass_Preferred
The Dog High mobility group protein B1 (HMGB1) ELISA Kit allows for quantitative determination of high-mobility group box 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Dog. Detection Range: 0.312 ng/mL-20 ng/mL. Sensitivity: 0.078 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q6YKA4. Restriction: For Research Use Only.
Catalog No. 50-149-7562