Learn More
Biomatik Corporation TOP2B Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50597
Encompass_Preferred
TOP2B Colorimetric Cell-Based ELISA Kit for detecting TOP2B in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q02880. Gene ID: 7155. Target Synonyms: DNA topoisomerase 2-beta; DNA topoisomerase II beta isozyme; TOP2B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0831