Learn More
Biomatik Corporation Tubulin γ Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50608
Encompass_Preferred
Tubulin γ Colorimetric Cell-Based ELISA Kit for detecting Tubulin γ in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P23258. Gene ID: 7283. Target Synonyms: Tubulin gamma-1 chain; Gamma-1-tubulin; Gamma-tubulin complex component 1; GCP-1; TUBG1; TUBG. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0841