Learn More
Biomatik Corporation BACH1 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50652
Encompass_Preferred
BACH1 Colorimetric Cell-Based ELISA for detecting BACH1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O14867. Gene ID: 571. Target Synonyms: BAC1; BTB and CNC homolog 1; HA2303; Transcription regulator protein BACH1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0878