Learn More
Biomatik Corporation TRI18 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50656
Encompass_Preferred
TRI18 Colorimetric Cell-Based ELISA for detecting TRI18 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O15344. Gene ID: 4281. Target Synonyms: Midline-1; Tripartite motif-containing protein 18; Putative transcription factor XPRF; Midin; RING finger protein 59; Midline 1 RING finger protein; MID1; FXY; RNF59; TRIM18; XPRF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0882