Learn More
Biomatik Corporation TAF5L Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50680
Encompass_Preferred
TAF5L Colorimetric Cell-Based ELISA for detecting TAF5L in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O75529. Gene ID: 27097. Target Synonyms: TAF5-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 5L; PCAF-associated factor 65 beta; PAF65-beta; TAF5L; PAF65B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0906