Learn More
Biomatik Corporation FIR Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50685
Encompass_Preferred
FIR Colorimetric Cell-Based ELISA for detecting FIR in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O94887. Gene ID: 9855. Target Synonyms: FERM; RhoGEF and pleckstrin domain protein 2; FGD1-related Cdc42-GEF; FIR; FRG; PLEKHC3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0911