Learn More
Biomatik Corporation ZNF265 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50688
Encompass_Preferred
ZNF265 Colorimetric Cell-Based ELISA for detecting ZNF265 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O95218. Gene ID: 9406. Target Synonyms: Z265; ZIS; ZN265; Zinc finger protein 265; Zinc finger; splicing. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0914