Learn More
Biomatik Corporation MBD3 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50694
Encompass_Preferred
MBD3 Colorimetric Cell-Based ELISA for detecting MBD3 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O95983. Gene ID: 53615. Target Synonyms: Methyl-CpG-binding domain protein 3; Methyl-CpG-binding protein MBD3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0920