Learn More
Biomatik Corporation HEXB Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50700
Encompass_Preferred
HEXB Colorimetric Cell-Based ELISA for detecting HEXB in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P07686. Gene ID: 3074. Target Synonyms: beta-hexosaminidase beta; beta-N-acetylhexosaminidase; cervical cancer proto-oncogene 7; HCC-7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0926