Learn More
Biomatik Corporation CEBPB Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50712
Encompass_Preferred
CEBPB Colorimetric Cell-Based ELISA for detecting CEBPB in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P17676. Gene ID: 1051. Target Synonyms: AGP/EBP; C/EBP beta; C/EBP-related protein 2; CCAAT/enhancer binding protein beta; CEBPB; CRP2; IL-6DBP; Interleukin-6- dependent binding protein; LAP; Liver-enriched transcriptional activator; Nuclear factor NF-IL6; SF-B; SFB; Silencer factor B; TCF5; Transcription factor 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0938