Learn More
Biomatik Corporation EGR1/2 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50714
Encompass_Preferred
EGR1/2 Colorimetric Cell-Based ELISA for detecting EGR1/2 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P18146; P11161. Gene ID: 1958; 1959. Target Synonyms: AT225; EGR-1; Early growth response protein 1; KROX-24; Krox-24 protein; NGFI-A; Nerve growth factor-induced protein A; Transcription factor ETR103; ZIF268; ZNF225; Zinc finger protein 225. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0940