Learn More
Biomatik Corporation DGKA Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50723
Encompass_Preferred
DGKA Colorimetric Cell-Based ELISA for detecting DGKA in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P23743. Gene ID: 1606. Target Synonyms: Diacylglycerol kinase alpha; DAG kinase alpha; Diglyceride kinase alpha; DGK-alpha; 80 kDa diacylglycerol kinase; DGKA; DAGK; DAGK1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0949