Learn More
Biomatik Corporation Dog von Willebrand Factor, vWF ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC32130
Encompass_Preferred
The Dog von Willebrand Factor, vWF ELISA Kit allows for quantitative determination of von Willebrand factor in serum, plasma and tissue homogenates. Species Reactivity: Dog. Detection Range: 0.625 ng/mL-40 ng/mL. Sensitivity: 0.32 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q28295. Restriction: For Research Use Only.
Catalog No. 50-149-7569