Learn More
Biomatik Corporation DGKE Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50771
Encompass_Preferred
DGKE Colorimetric Cell-Based ELISA for detecting DGKE in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P52429. Gene ID: 8526. Target Synonyms: Diacylglycerol kinase epsilon; Diglyceride kinase epsilon; DGK-epsilon; DAG kinase epsilon; DGKE; DAGK5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0995