Learn More
Biomatik Corporation RBM5 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50775
Encompass_Preferred
RBM5 Colorimetric Cell-Based ELISA for detecting RBM5 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P52756. Gene ID: 10181. Target Synonyms: G15; H37; LUCA15; RMB5; RNA binding motif protein 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0999