Learn More
Biomatik Corporation CDKL1 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50798
Encompass_Preferred
CDKL1 Colorimetric Cell-Based ELISA for detecting CDKL1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q00532. Gene ID: 8814. Target Synonyms: Cyclin-dependent kinase-like 1; Serine/threonine-protein kinase KKIALRE; Protein kinase p42 KKIALRE; CDKL1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1021