Learn More
Biomatik Corporation CSTF1 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50808
Encompass_Preferred
CSTF1 Colorimetric Cell-Based ELISA for detecting CSTF1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q05048. Gene ID: 1477. Target Synonyms: Cleavage stimulation factor subunit 1; CF-1 50 kDa subunit; CSTF 50 kDa subunit; CSTF1; Cleavage stimulation factor 50 kDa subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1030