Learn More
Biomatik Corporation TNFC Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50810
Encompass_Preferred
TNFC Colorimetric Cell-Based ELISA for detecting TNFC in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q06643. Gene ID: 4050. Target Synonyms: Lymphotoxin-beta; LT-beta; Tumor necrosis factor C; TNF-C; Tumor necrosis factor ligand superfamily member 3; LTB; TNFC; TNFSF3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1032