Learn More
Biomatik Corporation SFRS4 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50812
Encompass_Preferred
SFRS4 Colorimetric Cell-Based ELISA for detecting SFRS4 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q08170. Gene ID: 6429. Target Synonyms: Serine/arginine-rich splicing factor 4; Pre-mRNA-splicing factor SRP75; SRP001LB; SRSF4; SFRS4; SRP75. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1034