Learn More
Biomatik Corporation NCBP1 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50814
Encompass_Preferred
NCBP1 Colorimetric Cell-Based ELISA for detecting NCBP1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q09161. Gene ID: 4686. Target Synonyms: Nuclear cap-binding protein subunit 1; 80 kDa nuclear cap-binding protein; CBP80; NCBP 80 kDa subunit ; NCBP1 ; CBP80; NCBP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1036