Learn More
Biomatik Corporation Myelodysplasia Syndrome 1 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50831
Encompass_Preferred
Myelodysplasia Syndrome 1 Colorimetric Cell-Based ELISA for detecting Myelodysplasia Syndrome 1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13465. Gene ID: 4197. Target Synonyms: MDS1 and EVI1 complex locus protein MDS1; Myelodysplasia syndrome 1 protein; Myelodysplasia syndrome-associated protein 1; MECOM; MDS1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1052