Learn More
Biomatik Corporation GIT2 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50838
Encompass_Preferred
GIT2 Colorimetric Cell-Based ELISA for detecting GIT2 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q14161. Gene ID: 9815. Target Synonyms: ARF GTPase-activating protein GIT2; G protein-coupled receptor kinase-interactor 2; GRK-interacting protein 2; Cool-interacting tyrosine-Phospho protein 2; CAT2; CAT-2; GIT2; KIAA0148. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1059