Learn More
Biomatik Corporation CDK10 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50850
Encompass_Preferred
CDK10 Colorimetric Cell-Based ELISA for detecting CDK10 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15131. Gene ID: 8558. Target Synonyms: Cell division protein kinase 10; PISSLRE; Cdk10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1071