Learn More
Biomatik Corporation TAF1A Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50855
Encompass_Preferred
TAF1A Colorimetric Cell-Based ELISA for detecting TAF1A in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15573. Gene ID: 9015. Target Synonyms: TATA box-binding protein-associated factor RNA polymerase I subunit A; TATA box-binding protein-associated factor 1A; TBP-associated factor 1A; TBP-associated factor RNA polymerase I 48 kDa; TAFI48; Transcription factor SL1; TAF1A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1075