Learn More
Biomatik Corporation ADCK5 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50876
Encompass_Preferred
ADCK5 Colorimetric Cell-Based ELISA for detecting ADCK5 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q3MIX3. Gene ID: 203054. Target Synonyms: Uncharacterized aarF domain-containing protein kinase 5; ADCK5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1095