Learn More
Biomatik Corporation TTC23 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50885
Encompass_Preferred
TTC23 Colorimetric Cell-Based ELISA for detecting TTC23 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q5W5X9. Gene ID: 64927. Target Synonyms: TPR repeat protein 23; Cervical cancer proto-oncogene 8 protein; HCC-8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1104