Learn More
Biomatik Corporation SMRC2 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50911
Encompass_Preferred
SMRC2 Colorimetric Cell-Based ELISA for detecting SMRC2 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q8TAQ2. Gene ID: 6601. Target Synonyms: SWI/SNF complex 170 kDa subunit; BRG1-associated factor 170. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1130