Learn More
Biomatik Corporation ZNF785 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50917
Encompass_Preferred
ZNF785 Colorimetric Cell-Based ELISA for detecting ZNF785 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: A8K8V0. Gene ID: 146540. Target Synonyms: Zinc finger protein 785; ZNF785. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1136