Learn More
Biomatik Corporation Cytochrome P450 7B1 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50954
Encompass_Preferred
Cytochrome P450 7B1 Colorimetric Cell-Based ELISA for detecting Cytochrome P450 7B1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O75881. Gene ID: 9420. Target Synonyms: Cytochrome P450 7B1; 25-hydroxycholesterol 7-alpha-hydroxylase; Oxysterol 7-alpha-hydroxylase; CYP7B1; CP7B1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1172