Learn More
Biomatik Corporation PRKAG1/2/3 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50782
Encompass_Preferred
PRKAG1/2/3 Colorimetric Cell-Based ELISA for detecting PRKAG1/2/3 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P54619; Q9UGJ0; Q9UGI9. Gene ID: 5571; 51422; 53632;. Target Synonyms: 5'-AMP-activated protein kinase subunit gamma-1; AMPK gamma-1 chain; AMPKg. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1006