Learn More
Biomatik Corporation PITX1 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50791
Encompass_Preferred
PITX1 Colorimetric Cell-Based ELISA for detecting PITX1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P78337. Gene ID: 5307. Target Synonyms: Pituitary homeobox 1; Hindlimb-expressed homeobox protein backfoot; PITX1; BFT; PTX1; Homeobox protein PITX1; Paired-like homeodomain transcription factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1014