Learn More
Biomatik Corporation TNFRSF9 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50811
Encompass_Preferred
TNFRSF9 Colorimetric Cell-Based ELISA for detecting TNFRSF9 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q07011. Gene ID: 3604. Target Synonyms: Tumor necrosis factor receptor superfamily member 9; T-cell antigen 4-1BB homolog; T-cell antigen ILA; CD137 antigen; CDw137; TNFRSF9; CD137; ILA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1033