Learn More
Biomatik Corporation TCOF1 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50828
Encompass_Preferred
TCOF1 Colorimetric Cell-Based ELISA for detecting TCOF1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13428. Gene ID: 6949. Target Synonyms: Treacle protein; Treacher Collins syndrome protein; TCOF1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1049