Learn More
Biomatik Corporation GK3 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50841
Encompass_Preferred
GK3 Colorimetric Cell-Based ELISA for detecting GK3 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q14409; P32189. Gene ID: 2713. Target Synonyms: Putative glycerol kinase 3; GK 3; Glycerokinase 3; glycerol 3-phosphotransferase 3; Glycerol kinase testis specific 1 ; GK3P ; GKP3; GKTB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1062