Learn More
Biomatik Corporation MELK Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50843
Encompass_Preferred
MELK Colorimetric Cell-Based ELISA for detecting MELK in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q14680. Gene ID: 9833. Target Synonyms: Maternal embryonic leucine zipper kinase; hMELK; Protein kinase PK38; hPK38; MELK; KIAA0175. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1064