Learn More
Biomatik Corporation FAKD3 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50846
Encompass_Preferred
FAKD3 Colorimetric Cell-Based ELISA for detecting FAKD3 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q14CZ7. Gene ID: 79072. Target Synonyms: FAST kinase domain-containing protein 3; FASTKD3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1067