Learn More
Biomatik Corporation TSN Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50856
Encompass_Preferred
TSN Colorimetric Cell-Based ELISA for detecting TSN in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15631. Gene ID: 7247. Target Synonyms: Translin; TSN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1076