Learn More
Biomatik Corporation ZNF174 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50860
Encompass_Preferred
ZNF174 Colorimetric Cell-Based ELISA for detecting ZNF174 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15697. Gene ID: 7727. Target Synonyms: AW-1; Zinc finger and SCAN domain-containing protein 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1080