Learn More
Biomatik Corporation NAB2 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50862
Encompass_Preferred
NAB2 Colorimetric Cell-Based ELISA for detecting NAB2 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15742. Gene ID: 4665. Target Synonyms: EGR-1-binding protein 2; MADER; Melanoma- associated delayed early response protein; NGFI-A-binding protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1082