Learn More
Biomatik Corporation AKAP14 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50896
Encompass_Preferred
AKAP14 Colorimetric Cell-Based ELISA for detecting AKAP14 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q86UN6. Gene ID: 158798. Target Synonyms: A-kinase anchor protein 14; A-kinase anchor protein 14; A-kinase anchor protein 28 kDa; AKAP14; AKAP28. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1115