Learn More
Biomatik Corporation TRIM59 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50899
Encompass_Preferred
TRIM59 Colorimetric Cell-Based ELISA for detecting TRIM59 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q8IWR1. Gene ID: 286827. Target Synonyms: Tumor suppressor TSBF-1; RING finger protein 104. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1118