Learn More
Biomatik Corporation MAP4K6 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50903
Encompass_Preferred
MAP4K6 Colorimetric Cell-Based ELISA for detecting MAP4K6 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q8N4C8. Gene ID: 50488. Target Synonyms: Mitogen-activated protein kinase kinase kinase kinase 6; MAPK/ERK kinase kinase kinase 6; MEK kinase kinase 6; MEKKK 6; Misshapen/NIK-related kinase; GCK family kinase MiNK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1122