Learn More
Biomatik Corporation LMO3 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50910
Encompass_Preferred
LMO3 Colorimetric Cell-Based ELISA for detecting LMO3 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q8TAP4. Gene ID: 55885. Target Synonyms: LIM domain only protein 3; Neuronal-specific transcription factor DAT1; Rhombotin-3; LMO3; RBTN3; RBTNL2; RHOM3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1129