Learn More
Biomatik Corporation Cytochrome P450 26A1 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50940
Encompass_Preferred
Cytochrome P450 26A1 Colorimetric Cell-Based ELISA for detecting Cytochrome P450 26A1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O43174. Gene ID: 1592. Target Synonyms: Cytochrome P450 26A1; Retinoic acid-metabolizing cytochrome; P450 retinoic acid-inactivating 1; P450RAI; hP450RAI; Retinoic acid 4-hydroxylase; CYP26; P450RAI1; CP26A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1158