Learn More
Biomatik Corporation Cytochrome P450 2B6 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50942
Encompass_Preferred
Cytochrome P450 2B6 Colorimetric Cell-Based ELISA for detecting Cytochrome P450 2B6 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P20813. Gene ID: 1555. Target Synonyms: CP2B6; CPB1; CYP2B-1; CYP2B6; CYPIIB1; CYPIIB6; Cytochrome P450 2B1; Cytochrome P450 2B6; P450 IIB1; P450-B; P450-LM2; P450-PB1; P450-PB2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1160