Learn More
Biomatik Corporation CNTF (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51825
Encompass_Preferred
CNTF (Human) Sandwich ELISA Kit for detecting CNTF in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 47-3000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P26441. Gene ID: 1270. Target Synonyms: Ciliary neurotrophic factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1812