Learn More
Biomatik Corporation IL-12 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51841
Encompass_Preferred
IL-12 (Human) Sandwich ELISA Kit for detecting IL-12 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P29459; P29460. Gene ID: 3592; 3593. Target Synonyms: Interleukin-12 subunit alpha/beta; Cytotoxic lymphocyte maturation factor; NK cell stimulatory factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1818