Learn More
Biomatik Corporation IL-17F (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51845
Encompass_Preferred
IL-17F (Human) Sandwich ELISA Kit for detecting IL-17F in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q96PD4. Gene ID: 112744. Target Synonyms: Interleukin-17F; Cytokine ML-1; Interleukin-24. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1821