Learn More
Biomatik Corporation IL-20 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51849
Encompass_Preferred
IL-20 (Human) Sandwich ELISA Kit for detecting IL-20 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 16-4000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9NYY1. Gene ID: 50604. Target Synonyms: Interleukin-20; Cytokine Zcyto10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1825